Anti-SEM1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SEM1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77799-516
- 77799-515
- Antigen Symbol:
- SEM1
- SEM1
- Antigen Name:
- 26S proteasome complex subunit SEM1 protein (1-70AA)
- 26S proteasome complex subunit SEM1 protein (1-70AA)
- Antigen Synonyms:
- SEM1|C7orf76|DSS1|SHFDG1|SHFM1|26S proteasome complex subunit SEM1|26S proteasome complex subunit DSS1|Deleted in split hand/split foot protein 1|Split hand/foot deleted protein 1|Split hand/foot malformation type 1 protein
- SEM1|C7orf76|DSS1|SHFDG1|SHFM1|26S proteasome complex subunit SEM1|26S proteasome complex subunit DSS1|Deleted in split hand/split foot protein 1|Split hand/foot deleted protein 1|Split hand/foot malformation type 1 protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P60896
- P60896
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice