Anti-SLC27A5 Rabbit Polyclonal Antibody
77839-618
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antigen Name:Synthesized peptide derived from internal of SLC27A5
- Antigen Symbol:SLC27A5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q9Y2P5
- Antigen Synonyms:ACSB|ACSVL6|BA CoA ligase|BA-CoA ligase|BACS|BAL|Bile acid CoA ligase|Bile acid-CoA ligase|Bile acyl CoA synthetase|Bile acyl-CoA synthetase|Cholate CoA ligase|Cholate--CoA ligase|FACVL3|FATP 5|FATP-5|FATP5|Fatty acid Coenzyme A ligase very long chain 3|Fatty acid transport protein 5|Fatty-acid-coenzyme A ligase|FLJ22987|S27A5_HUMAN|Slc27a5|Solute carrier family 27 (fatty acid transporter) member 5|Solute carrier family 27 member 5|Very long chain acyl CoA synthetase homolog 2|Very long chain acyl CoA synthetase related protein|very long-chain 3|Very long-chain acyl-CoA synthetase homolog 2|Very long-chain acyl-CoA synthetase-related protein|VLACS related|VLACS-related|VLACSR|VLCS H2|VLCS-H2|VLCSH2
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:3000, IHC: 1:50 to 1:100, IF: 1:100 to 1:500
Type:
Antigen: SLC27A5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human