Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-ANKMY1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
 
undefined
Anti-ANKMY1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product

 

  • Catalog No:
  • 77842-612
  • 77842-613
  • Antigen Symbol:
  • ANKMY1
  • ANKMY1
  • Antigen Name:
  • Ankyrin repeat and MYND domain-containing protein 1 protein (1-220AA)
  • Ankyrin repeat and MYND domain-containing protein 1 protein (1-220AA)
  • Antigen Synonyms:
  • ANKMY1|TSAL1|ZMYND13Ankyrin repeat and MYND domain-containing protein 1|Testis-specific ankyrin-like protein 1|Zinc finger MYND domain-containing protein 13
  • ANKMY1|TSAL1|ZMYND13Ankyrin repeat and MYND domain-containing protein 1|Testis-specific ankyrin-like protein 1|Zinc finger MYND domain-containing protein 13
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • HRP (Horseradish Peroxidase)
  • HRP (Horseradish Peroxidase)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9P2S6
  • Q9P2S6
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice