Anti-GLRX5 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: GLRX5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77831-889
- 77831-890
- Antigen Symbol:
- GLRX5
- GLRX5
- Antigen Name:
- Glutaredoxin-related protein 5, mitochondrial protein (32-157AA)
- Glutaredoxin-related protein 5, mitochondrial protein (32-157AA)
- Antigen Synonyms:
- C14orf87|Chromosome 14 open reading frame 87|FLB4739|GLRX 5|Glrx5|_HUMAN|Glutaredoxin 5 homolog|Glutaredoxin related protein 5|Glutaredoxin-related protein 5|Glutaredoxin5|GRX5|MGC14129|mitochondrial|Monothiol glutaredoxin-5|PRO1238
- C14orf87|Chromosome 14 open reading frame 87|FLB4739|GLRX 5|Glrx5|_HUMAN|Glutaredoxin 5 homolog|Glutaredoxin related protein 5|Glutaredoxin-related protein 5|Glutaredoxin5|GRX5|MGC14129|mitochondrial|Monothiol glutaredoxin-5|PRO1238
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86SX6
- Q86SX6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice