Anti-MYLK3 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: MYLK3
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77826-060
- 77826-061
- Antigen Symbol:
- MYLK3
- MYLK3
- Antigen Name:
- Myosin light chain kinase 3 protein (1-240AA)
- Myosin light chain kinase 3 protein (1-240AA)
- Antigen Synonyms:
- caMLCK| cardiac MLCK| Cardiac MyBP C associated Ca/CaM kinase| Cardiac-MyBP-C-associated Ca/CaM kinase| MLC kinase| MLCK| MLCK2| Mylk3| MYLK3_HUMAN| Myosin light chain kinase 3| Putative myosin light chain kinase 3
- caMLCK| cardiac MLCK| Cardiac MyBP C associated Ca/CaM kinase| Cardiac-MyBP-C-associated Ca/CaM kinase| MLC kinase| MLCK| MLCK2| Mylk3| MYLK3_HUMAN| Myosin light chain kinase 3| Putative myosin light chain kinase 3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q32MK0
- Q32MK0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice