Anti-TSPAN6 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TSPAN6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77803-639
- 77811-639
- Antigen Symbol:
- TSPAN6
- TSPAN6
- Antigen Name:
- Tetraspanin-6 protein (115-208AA)
- Tetraspanin-6 protein (115-208AA)
- Antigen Synonyms:
- 6720473L21Rik|A15 homolog|AI316786|DKFZp469J2433|MGC117923|MGC133784|Putative NF kappa B activating protein 321|Putative NF-kappa-B-activating protein 321|T245|T245 protein|Tetraspan TM4SF|Tetraspanin 6|Tetraspanin TM4 D|Tetraspanin TM4-D|Tetraspanin TM4D|Tetraspanin-6|Tm4sf|TM4SF6|Transmembrane 4 superfamily member 6|TSN6_HUMAN|TSPAN 6|Tspan-6|TSPAN6|Tspan6 protein
- 6720473L21Rik|A15 homolog|AI316786|DKFZp469J2433|MGC117923|MGC133784|Putative NF kappa B activating protein 321|Putative NF-kappa-B-activating protein 321|T245|T245 protein|Tetraspan TM4SF|Tetraspanin 6|Tetraspanin TM4 D|Tetraspanin TM4-D|Tetraspanin TM4D|Tetraspanin-6|Tm4sf|TM4SF6|Transmembrane 4 superfamily member 6|TSN6_HUMAN|TSPAN 6|Tspan-6|TSPAN6|Tspan6 protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43657
- O43657
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice