Anti-USP14 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: USP14
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77804-119
- 77804-118
- Antigen Symbol:
- USP14
- USP14
- Antigen Name:
- Ubiquitin carboxyl-terminal hydrolase 14 protein (1-494AA)
- Ubiquitin carboxyl-terminal hydrolase 14 protein (1-494AA)
- Antigen Synonyms:
- Deubiquitinating enzyme 14|TGT|tRNA guanine transglycosylase 60 kD subunit|tRNA guanine transglycosylase|Ubiquitin carboxyl terminal hydrolase 14|Ubiquitin carboxyl-terminal hydrolase 14|Ubiquitin specific peptidase 14|Ubiquitin specific processing protease 14|Ubiquitin specific protease 14|Ubiquitin thiolesterase 14|Ubiquitin-specific-processing protease 14|UBP14_HUMAN|USP 14|USP14
- Deubiquitinating enzyme 14|TGT|tRNA guanine transglycosylase 60 kD subunit|tRNA guanine transglycosylase|Ubiquitin carboxyl terminal hydrolase 14|Ubiquitin carboxyl-terminal hydrolase 14|Ubiquitin specific peptidase 14|Ubiquitin specific processing protease 14|Ubiquitin specific protease 14|Ubiquitin thiolesterase 14|Ubiquitin-specific-processing protease 14|UBP14_HUMAN|USP 14|USP14
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P54578
- P54578
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice