Anti-RTL8A Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RTL8A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77838-047
- 77838-048
- Antigen Symbol:
- RTL8A
- RTL8A
- Antigen Name:
- Retrotransposon Gag-like protein 8A protein (1-113AA)
- Retrotransposon Gag-like protein 8A protein (1-113AA)
- Antigen Synonyms:
- CXX1B|FAM127B|MAR8A|Retrotransposon Gag-like protein 8A|Mammalian retrotransposon derived protein 8A
- CXX1B|FAM127B|MAR8A|Retrotransposon Gag-like protein 8A|Mammalian retrotransposon derived protein 8A
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BWD3
- Q9BWD3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice