Anti-USP25 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: USP25
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77841-404
- 77841-405
- Antigen Symbol:
- USP25
- USP25
- Antigen Name:
- Ubiquitin carboxyl-terminal hydrolase 25 protein (348-606AA)
- Ubiquitin carboxyl-terminal hydrolase 25 protein (348-606AA)
- Antigen Synonyms:
- Deubiquitinating enzyme 25|Ubiquitin carboxyl-terminal hydrolase 25|Ubiquitin specific peptidase 25|ubiquitin specific protease 25|ubiquitin specific protease USP25|Ubiquitin thiolesterase 25|Ubiquitin-specific-processing protease 25|UBP25_HUMAN|USP on chromosome 21|USP21|USP25
- Deubiquitinating enzyme 25|Ubiquitin carboxyl-terminal hydrolase 25|Ubiquitin specific peptidase 25|ubiquitin specific protease 25|ubiquitin specific protease USP25|Ubiquitin thiolesterase 25|Ubiquitin-specific-processing protease 25|UBP25_HUMAN|USP on chromosome 21|USP21|USP25
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UHP3
- Q9UHP3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice