Anti-PRKAB1 Rabbit Polyclonal Antibody
77827-226
New Product
- Antigen Name:Synthesized peptide derived from N-terminal of PRKAB1.
- Antigen Symbol:PRKAB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:liquid
- Gene ID:Q9Y478
- Antigen Synonyms:1300015D22Rik|5''-AMP-activated protein kinase subunit beta-1|5'-AMP-activated protein kinase beta-1 subunit|AAKB1_HUMAN|AMP-activated protein kinase beta subunit|AMP-ACTIVATED PROTEIN KINASE|NONCATALYTIC|BETA-1|AMP-activated|noncatalytic|beta-1|AMPK|AMPK beta 1 chain|AMPK subunit beta-1|AMPK-BETA-1|AMPKb|AU021155|E430008F22|HAMPKb|MGC17785|PRKAB1|Protein kinase AMP activatedNon catalytic subunit beta 1|protein kinase|beta 1Non-catalytic subunit
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: PRKAB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat