Anti-RASA2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000
Type:
Antigen: RASA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77820-524
- 77820-523
- Antigen Symbol:
- RASA2
- RASA2
- Antigen Name:
- Synthetic peptide of RASA2
- Synthetic peptide of RASA2
- Antigen Synonyms:
- GAP1m|GTPase activating protein of RAS|Ras GTPase activating protein 2|Ras GTPase-activating protein 2|RAS p21 protein activator 2|Rasa2|RASA2_HUMAN
- GAP1m|GTPase activating protein of RAS|Ras GTPase activating protein 2|Ras GTPase-activating protein 2|RAS p21 protein activator 2|Rasa2|RASA2_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15283
- Q15283
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice