5-Methoxyindole ≥99.0%
About this item
CAS Number: 1006-94-6
MDL Number: MFCD00005674
Molecular Formula: C9H9NO
Molecular Weight: 147.18
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 178
Melting point (°C): 55
Storage Temperature: 0-10°C
- CAS Number:1006-94-6
- Formula:C₉H₉NO
- MDL Number:MFCD00005674
- Molecular Weight:147.18 g/mol
- Boiling Pt:178 °C
- Melting Pt:55 °C
- Storage Temperature:Refrigerator
- EINECS:213-745-3
3 Options Available Below
Product Details & Documents
CAS Number: 1006-94-6
MDL Number: MFCD00005674
Molecular Formula: C9H9NO
Molecular Weight: 147.18
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 178
Melting point (°C): 55
Storage Temperature: 0-10°C
- CAS Number:1006-94-6
- Formula:C₉H₉NO
- MDL Number:MFCD00005674
- Molecular Weight:147.18 g/mol
- Boiling Pt:178 °C
- Melting Pt:55 °C
- Storage Temperature:Refrigerator
- EINECS:213-745-3
Specifications for Product Family
Specification Test Results
- Purity/Analysis Method:>99.0% (GC)
Product Family Options
Product Information
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



