Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
4-Methylanisole ≥98.0%

4-Methylanisole ≥98.0%

Supplier:   TCI America
4-Methylanisole ≥98.0%
Supplier:  TCI America
Certificates
4-Methylanisole ≥98.0%

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 104-93-8
MDL Number: MFCD00008413
Molecular Formula: C8H10O
Molecular Weight: 122.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 175
Melting point (°C): -50
Flash Point (°C): 59
Specific Gravity (20/20): 0.97
Lambda max.: 279 nm (MeOH)

Other Product Names / Synonyms: p-Cresol Methyl Ether, 4-Methylanisole

Synonyms:  p-Methylanisole
  • CAS Number:104-93-8
  • Formula:C₈H₁₀O
  • MDL Number:MFCD00008413
  • Molecular Weight:122.17 g/mol
  • Boiling Pt:175 °C
  • Melting Pt:–50 °C
  • Density:0.97 (20 °C)
  • Flash Pt:59 °C
  • Storage Temperature:Ambient
  • EINECS:203-253-7
  • UN:3271
  • ADR:3,III

Documents

Certificates
Search by Lot Number
Specifications for Product Family

Specification Test Results

  • Purity/Analysis Method:
    >98.0% (GC)
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 25 mL
    Log in for availability
    Log in for availability
    Each (25ml)
    Each (25ml) - $18.00
    $18.00
    Each (25ml)
       $18.00

    Supplier Number: M0149-25ML
  • 500 mL
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $60.00
    $60.00
    Each (500ml)
       $60.00

    Supplier Number: M0149-500ML

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.