2-Propanol ≥99.5%
Product Details & Documents
[for HPLC Solvent]
CAS Number: 67-63-0
MDL Number: MFCD00011674
Molecular Formula: C3H8O
Molecular Weight: 60.10
Purity/Analysis Method: >99.5% (GC)
Form: Clear Liquid
Boiling point (°C): 82
Flash Point (°C): 15
Specific Gravity (20/20): 0.79
Product Categories: Analytical Science, Solvents for HPLC
Other Product Names / Synonyms: Isopropanol, 2-Propanol, IPA
- CAS Number:67-63-0
- Formula:(CH₃)₂CHOH
- MDL Number:MFCD00011674
- Molecular Weight:60.1 g/mol
- Merck Index:13,05228
- Boiling Pt:82 °C
- Melting Pt:–89 °C
- Density:0.79 (20 °C)
- Flash Pt:15 °C
- Storage Temperature:Ambient
- EINECS:200-661-7
- UN:1219
- ADR:3,II
- REACH:01-2119457558-25
Specifications
Specification Test Results
- Purity/Analysis Method:>99.5% (GC)
Specifications
- Size:500 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



