2-Hexanone ≥98.0%
About this item
CAS Number: 591-78-6
MDL Number: MFCD00009482
Molecular Formula: C6H12O
Molecular Weight: 100.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 127
Melting point (°C): -57
Flash Point (°C): 23
Specific Gravity (20/20): 0.81
Other Product Names / Synonyms: Butyl Methyl Ketone
- CAS Number:591-78-6
- Formula:C₆H₁₂O
- MDL Number:MFCD00009482
- Molecular Weight:100.16 g/mol
- Merck Index:13,06060
- Boiling Pt:127 °C
- Melting Pt:–57 °C
- Density:0.81 (20 °C)
- Flash Pt:23 °C
- Storage Temperature:Ambient
- EINECS:209-731-1
- UN:1224
- ADR:3,III
2 Options Available Below
Product Details & Documents
CAS Number: 591-78-6
MDL Number: MFCD00009482
Molecular Formula: C6H12O
Molecular Weight: 100.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 127
Melting point (°C): -57
Flash Point (°C): 23
Specific Gravity (20/20): 0.81
Other Product Names / Synonyms: Butyl Methyl Ketone
- CAS Number:591-78-6
- Formula:C₆H₁₂O
- MDL Number:MFCD00009482
- Molecular Weight:100.16 g/mol
- Merck Index:13,06060
- Boiling Pt:127 °C
- Melting Pt:–57 °C
- Density:0.81 (20 °C)
- Flash Pt:23 °C
- Storage Temperature:Ambient
- EINECS:209-731-1
- UN:1224
- ADR:3,III
Documents
Certificates
Search by Lot Numberghs_H0114
SDS
VWR GHS Compliance Letter (US)
Additional Product Details
Specifications for Product Family
Specification Test Results
- Purity/Analysis Method:>98.0% (GC)
Product Family Options
Product Information
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



