Trityl chloride ≥98.0%
About this item
CAS Number: 76-83-5
MDL Number: MFCD00000813
Molecular Formula: C19H15Cl
Molecular Weight: 278.78
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 310
Melting point (°C): 113
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Nucleic Acid Synthesis
Other Product Names / Synonyms: Chlorotriphenylmethane, Triphenylchloromethane, Triphenylmethyl Chloride
- CAS Number:76-83-5
- Formula:C₁₉H₁₅Cl
- MDL Number:MFCD00000813
- Molecular Weight:278.78 g/mol
- Boiling Pt:310 °C
- Melting Pt:113 °C
- Density:1.25 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-986-4
- UN:3261
- ADR:8,II
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
CAS Number: 76-83-5
MDL Number: MFCD00000813
Molecular Formula: C19H15Cl
Molecular Weight: 278.78
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 310
Melting point (°C): 113
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Nucleic Acid Synthesis
Other Product Names / Synonyms: Chlorotriphenylmethane, Triphenylchloromethane, Triphenylmethyl Chloride
- CAS Number:76-83-5
- Formula:C₁₉H₁₅Cl
- MDL Number:MFCD00000813
- Molecular Weight:278.78 g/mol
- Boiling Pt:310 °C
- Melting Pt:113 °C
- Density:1.25 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-986-4
- UN:3261
- ADR:8,II
Documents
Certificates
Search by Lot Numberghs_C0308
SDS
VWR GHS Compliance Letter (US)
Additional Product Details
Specifications for Product Family
Specification Test Results
- Purity/Analysis Method:>98.0% (T)
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: C0308-25GItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Supplier Number: C0308-100GItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



