Order Entry
Puerto Rico
ContactUsLinkComponent
Clorofene ≥97.0%
Clorofene ≥97.0%
Catalog # TCB0812-025G
Supplier:  TCI America
CAS Number:  
Clorofene ≥97.0%
Catalog # TCB0812-025G
Supplier:  TCI America
Supplier Number:  B0812-25G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    25 g
  • Cat. No.:
    TCB0812-025G
  • Supplier no.:
    B0812-25G

Specifications

Specification Test Results

  • Purity/Analysis Method:
    >97.0% (GC)

Specification Test Results

About this item

 :
 :218.68 g/mol
Merck Index:13,02433
Boiling Pt:162 °C
Boiling Pt:162 °C
Melting Pt:47 °C
Density:1.22 g/cm³ (20 °C)
Flash Pt:185 °C
EINECS:204-385-8
CAS Number:120-32-1
Formula:C₁₃H₁₁ClO
MDL Number:MFCD00020140
Flash Pt:185 °C