2-Hydroxyethyl macrylate ≥95.0% stabilized
Catalog # TCA0743-25ML
Supplier: TCI America
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Size:25 ml
- Cat. No.:TCA0743-25ML
- Supplier no.:A0743-25ML
Specifications
Specification Test Results
- Purity/Analysis Method:>95.0% (GC)
Specification Test Results
About this item
CAS Number:818-61-1
:
Formula:C₅H₈O₃
:116.12 g/mol
Boiling Pt:92 °C
Density:1.11 (20 °C)
Flash Pt:104 °C
EINECS:212-454-9
UN:2927
ADR:6.1,II
:Stabilised with Stabilised
MDL Number:MFCD00002865