


Ethylamine ≥70% in aqueous solution
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:75-04-7
:
Formula:C₂H₇N
MDL Number:MFCD00008160
:45.08
Merck Index:14,3762
Boiling Pt:∼ 38 °C
Melting Pt:–81 °C
Density:0.803
Flash Pt:–17 °C (1 °F)
Storage Temperature:Ambient
EINECS:200-834-7
UN:2270
ADR:3,II
:GHS02
:GHS05
:GHS06
Related Information
Certificates
Search by Lot NumberAlfa Aesar_Buy 3 Get 2 Complimentary items
Promotion Material
Organic Building Blocks and Intermediates
Application Notes
VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together