Order Entry
Puerto Rico
ContactUsLinkComponent
Ethanol denatured, anhydrous ≥90% for HPLC, for LC-MS (denatured with 5% isopropanol and 5% methanol)
Flammable
Health
Ethanol denatured, anhydrous ≥90% for HPLC, for LC-MS (denatured with 5% isopropanol and 5% methanol)
Catalog # AA44134-AE
CAS Number:  
Flammable
Health
Ethanol denatured, anhydrous ≥90% for HPLC, for LC-MS (denatured with 5% isopropanol and 5% methanol)
Catalog # AA44134-AE
Supplier Number:  044134-AE
CAS Number:  

Specifications

  • Size:
    100 ml
  • Cat. No.:
    AA44134-AE
  • Supplier no.:
    44134-AE

Specifications

Specification Test Results

  • Evaporation residue:
    5ppm max.
  • H2O:
    0.1% max.

Specification Test Results

About this item

 :
 :46.07 g/mol
Merck Index:14,3760
Boiling Pt:78 °C
Melting Pt:–90 °C
Density:0.789
Flash Pt:11 °C (52 °F)
EINECS:200-578-6
UN:1987
ADR:3,II
 :Packaged under argon in resealable ChemSeal™ bottles.
 :GHS02
 :GHS08
Storage Temperature:Ambient
CAS Number:64-17-5
Formula:H₃CCH₂OH
MDL Number:MFCD00003568