

Ethanol denatured, anhydrous ≥90% for spectrophotometry (denatured with 5% isopropanol and 5% methanol)
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:64-17-5
:
Formula:H₃CCH₂OH
MDL Number:MFCD00003568
:46.07 g/mol
Merck Index:14,3760
Boiling Pt:78 °C
Melting Pt:–90 °C
Density:0.789
Flash Pt:11 °C (52 °F)
Storage Temperature:Ambient
EINECS:200-578-6
UN:1987
ADR:3,II
:GHS02
:GHS08
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:EPA Safer chemical solvent (green circle rating); Chem21 solvent selection guide recommended by default and after discussion
Specifications
Specification Test Results
- Evaporation residue:5ppm max.
- H2O:0.1% max.
- Reagent alcohol:0.05% max.
Specification Test Results
Related Information
Certificates
Search by Lot NumberAlfa Aesar_Buy 3 Get 2 Complimentary items
Promotion Material
VWR GHS Compliance Letter (US)