Beryllium, Plasma Standard Solution, Specpure®, Be 1000µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Molecular Formula: Be4O(C2H3O2)6 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134032
Specifications
- MDL No.:MFCD00134032
Specifications
Related Information
Certificates
Search by Lot NumberAlfa Aesar_Buy 3 Get 2 Complimentary items
Promotion Material
VWR GHS Compliance Letter (US)