Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
3,4-Dimethoxyphenylglyoxal hydrate 98% (dry weight)

3,4-Dimethoxyphenylglyoxal hydrate 98% (dry weight)

Irritant
3,4-Dimethoxyphenylglyoxal hydrate 98% (dry weight)
Supplier:  
Certificates
3,4-Dimethoxyphenylglyoxal hydrate 98% (dry weight)
3,4-Dimethoxyphenylglyoxal hydrate 98% (dry weight)

0 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 163428-90-8 Molecular Formula: C10H10O4.xH2O Formula Weight: 194.19(anhy) Melting Point: 118-120° Beilstein Reference: 3092619 MDL No.: MFCD06800598

  • CAS Number:163428-90-8
  • Formula:C₁₀H₁₀O₄ xH₂O
  • MDL Number:MFCD06800598
  • Molecular Weight:194.19 g/mol (anhy)
  • Melting Pt:118…120 °C
  • Storage Temperature:Ambient

Documents

Product Family Options
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.