Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Hypericin 98%

Hypericin 98%
Certificates

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Inhibitor of protein kinase C; induces photosynthesized apoptosis

Purity: 98% CAS No.: 548-04-9 Molecular Formula: C30H16O8 Formula Weight: 504.44 Storage Temperature: -30°C to -10°C Melting Point: ca 320° dec. Beilstein Reference: 1917913 MDL No.: MFCD00016683

Synonyms:  1,3,4,6,8,13-Hexahydroxy-10,11-dimethylphenanthro[1,10,9,8-opqra]perylene-7,14-dione P-conformer
  • CAS Number:548-04-9
  • Formula:C₃₀H₁₆O₈
  • MDL Number:MFCD00016683
  • Molecular Weight:504.45 g/mol
  • Merck Index:14,4863
  • Melting Pt:∼ 320 °C (dec.)
  • Melting Pt:ca 320 °C (dec.)
  • Storage Temperature:Refrigerator
  • EINECS:208-941-0
Product Family Options

Product Information

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.