Order Entry
Puerto Rico
ContactUsLinkComponent
D-Glucose Dehydrogenase, Thermo Scientific
D-Glucose Dehydrogenase, Thermo Scientific
Catalog # AA45402-4I
CAS Number:  
D-Glucose Dehydrogenase, Thermo Scientific
Catalog # AA45402-4I
Supplier Number:  045402-4I
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • CAS No.:
    9028-53-9
  • Size:
    1 KU
  • MDL No.:
    MFCD00131181
  • Enzyme Name:
    D-Glucose Dehydrogenase
  • Cat. No.:
    AA45402-4I
  • Supplier no.:
    45402-4I

Specifications

About this item

Uses: diagnostics and cofactor regeneration of NAD(P)H in reductive processes.

Physical Form: Powder Appearance: Pale yellow MDL No.: MFCD00131181

Applications: Diagnostics and cofactor regeneration of NAD(P)H in reductive processes