Order Entry
Puerto Rico
ContactUsLinkComponent
Polystyrene standard, M.W. 290,000, Mw/Mn 1.06, Thermo Scientific
Polystyrene standard, M.W. 290,000, Mw/Mn 1.06, Thermo Scientific
Catalog # AA41937-03
CAS Number:  
Polystyrene standard, M.W. 290,000, Mw/Mn 1.06, Thermo Scientific
Catalog # AA41937-03
Supplier Number:  041937-03
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • CAS No.:
    9003-53-6
  • Size:
    1 g
  • MDL No.:
    MFCD00084450
  • Formula:
    (C₈H₈)n
  • Description:
    Polystyrene Standard
  • Physical Form:
    Powder
  • Odor/Scent:
    Odorless
  • Cat. No.:
    AA41937-03
  • Supplier no.:
    41937-03

Specifications

About this item

CAS No.: 9003-53-6
Physical Form: Powder
Odor: Odorless
MDL No.: MFCD00084450