Order Entry
Puerto Rico
ContactUsLinkComponent
Silicon dioxide ≥99.995% (metals basis), sputtering target, Diameter 50.8 mm (2.0 in), Thickness 3.18 mm (0.125 in)
Silicon dioxide ≥99.995% (metals basis), sputtering target, Diameter 50.8 mm (2.0 in), Thickness 3.18 mm (0.125 in)
Catalog # AA41096-KS
CAS Number:  
Silicon dioxide ≥99.995% (metals basis), sputtering target, Diameter 50.8 mm (2.0 in), Thickness 3.18 mm (0.125 in)
Catalog # AA41096-KS
Supplier Number:  041096-KS
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    1 each
  • Cat. No.:
    AA41096-KS
  • Supplier no.:
    41096-KS

Specifications

About this item

CAS Number:7631-86-9
 :
Formula:SiO₂
MDL Number:MFCD00011232
 :60.08 g/mol
Merck Index:14,08493
Boiling Pt:2230 °C (1013 hPa)
Melting Pt:1719 °C
Density:2.2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-545-4