Order Entry
Puerto Rico
ContactUsLinkComponent
Nickel (II) nitrate 1% Ni(NO3)2 in nitric acid 2%, Specpure®
Health
Corrosive
Nickel (II) nitrate 1% Ni(NO3)2 in nitric acid 2%, Specpure®
Catalog # AA39043-AE
CAS Number:  
Health
Corrosive
Nickel (II) nitrate 1% Ni(NO3)2 in nitric acid 2%, Specpure®
Catalog # AA39043-AE
Supplier Number:  039043-AE
CAS Number:  

Specifications

  • Size:
    100 ml
  • Cat. No.:
    AA39043-AE
  • Supplier no.:
    39043-AE

Specifications

About this item

 :
MDL Number:MFCD00011139
 :182.7
Storage Temperature:Ambient
UN:3264
ADR:8,III
 :GHS08
 :GHS05
CAS Number:13138-45-9
Formula:N₂NiO₆