Order Entry
Puerto Rico
ContactUsLinkComponent
Lanthanum(III) chloride 1% La (La2O3) in HCl, Specpure®
Corrosive
Lanthanum(III) chloride 1% La (La2O3) in HCl, Specpure®
Catalog # AA39036-AP
CAS Number:  
Corrosive
Lanthanum(III) chloride 1% La (La2O3) in HCl, Specpure®
Catalog # AA39036-AP
Supplier Number:  039036-AP
CAS Number:  

Specifications

  • Size:
    500 ml
  • Cat. No.:
    AA39036-AP
  • Supplier no.:
    39036-AP

Specifications

About this item

 :
MDL Number:MFCD00011068
 :245.26 g/mol
Storage Temperature:Ambient
EINECS:231-595-7
UN:3264
ADR:8,III
 :GHS05
CAS Number:10099-58-8
Formula:Cl₃La