To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Rhodopsin
- Antigen Symbol:RHO
- Clonality:Monoclonal
- Clone:1D4
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Bovine,Mouse
- Western Blot:Yes
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Antibody fraction
- Form:Liquid
- Gene ID:509933
- Antigen Synonyms:Opsin 2|Rhodopsin|rod pigment|OPN2|Opsin-2|CSNBAD1|RP4|RHO
- Storage Buffer:0.01 M HEPES, 0.15 M Sodium Chloride, pH 7.50.1 mg/ml Bovine Serum Albumin - IgG and Protease free, 50% (v/v) Glycerol
- Storage Temperature:–20 °C
- Shipping Temperature:Dry Ice
- Immunogen:Anti-Rhodopsin Monoclonal Antibody was produced in mouse by repeated immunizations with purified native bovine rhodopsin.
- Purification:Protein G purified
- Cat. No.:RL200-301-E29
- Supplier no.:200-301-E29
Specifications
About this item
Anti-Rhodopsin antibody is suitable for use in Western Blot and immunochemistry - IHC. Expect a band of approximately 39 kDa in size corresponding to the rhodopsin Proteins in Western blot in the appropriate cell lysate or extract.
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:100
Type: Primary
Antigen: RHO
Clonality: Monoclonal
Clone: 1D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Bovine