Anti-cells in xenographic research Mouse Polyclonal Antibody [clone: 235-1]
Catalog # RL209-301-D99
Supplier: Rockland Immunochemical
Specifications
- Antibody Type:Primary
- Antigen Name:cells in xenographic research
- Antigen Symbol:cells in xenographic research
- Clonality:Polyclonal
- Clone:235-1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Size:100 µL
- Cat. No.:RL209-301-D99
- Supplier no.:909-301-D99
Specifications
About this item
Type: Primary
Antigen: cells in xenographic research
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human