Elite Mini PAP Pen, Diagnostic BioSystems
Catalog # 95025-252
Supplier: Diagnostic Biosystems
CAS Number:
Specifications
- Description:Elite Mini PAP Pen
- Size:1 each
- Cat. No.:95025-252
- Supplier no.:K 042
Specifications
About this item
Elite mini PAP pen is a special marking pen that provides a thin film-like green-tinged hydrophobic barrier when a circle is drawn around a specimen on a slide. This water repellent barrier keeps staining reagents localized on the tissue sections, prevents mixing of reagents when differentially staining two sections on the same slide, and allows use of less reagents per section.
- Insoluble in alcohol and acetone
- Finer tip than the regular PAP pen
PAP Pen markings are insoluble in alcohol and acetone. It can be optionally removed with xylene after the staining procedure is complete. The pen is especially useful when working with multiple sections on a single slide because of its finer tip.