Anti-MECP2 Rabbit Polyclonal Antibody
Catalog # RL100-401-D71
Supplier: Rockland Immunochemical
Specifications
- Antibody Type:Primary
- Antigen Name:Methyl CPG Binding Protein 2
- Antigen Symbol:MECP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µL
- Cross Adsorption:No
- Format:Antiserum
- Form:Liquid
- Gene ID:4204
- Antigen Synonyms:RTS|MRX79|AUTSX3|MRX16|PPMX|RS|MRXS13|MRXSL|RTT
- Modification:phospho-Ser80
- Storage Buffer:0.01 M HEPES, 0.15 M Sodium Chloride, pH 7.50.1 mg/ml Bovine Serum Albumin - IgG and Protease free, 50% (v/v) Glycerol
- Storage Temperature:–20 °C
- Shipping Temperature:Dry Ice
- Immunogen:Anti-MeCP2 pS80 Antibody was produced in rabbit by repeated immunizations with MeCP Ser80 - Synthetic peptide corresponding to amino acid residues surrounding Ser 80 conjugated to KLH.
- Cat. No.:RL100-401-D71
- Supplier no.:100-401-D71
Specifications
About this item
Anti-MeCP2 pS80 (Rabbit) is suitable for use in Western Blotting and IHC. Anti-MeCP2 pS80 antibodies have been used successfully for IHS on mouse brain sections. Specific conditions for reactivity should be optimized by the end user.
- Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:100
Type: Primary
Antigen: MECP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Human