Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Buffers
Electrophoresis Buffers
Washing Buffers
Immuno Wash Buffer (10X), Diagnostic Biosystems
Immuno Wash Buffer (10X), Diagnostic Biosystems
Immuno Wash Buffer (10X), Diagnostic Biosystems
Catalog # 99990-062
Supplier:
Diagnostic Biosystems
CAS Number:
Expand image
Immuno Wash Buffer (10X), Diagnostic Biosystems
Catalog # 99990-062
Supplier:
Diagnostic Biosystems
Supplier Number:
K005
CAS Number:
Specifications
Description:
10X Immuno Wash Buffer
Size:
1 l
Cat. No.:
99990-062
Supplier no.:
K 005
Specifications
About this item
Product Details
Documentation
Product Details
Documentation