Anti-HSA Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # RL600-402-033
Supplier: Rockland Immunochemical
Specifications
- Antibody Type:Primary
- Antigen Symbol:HSA
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:1 mg
- Cross Adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Antigen Synonyms:Anti-Human Serum Albumin
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
- Storage Temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping Temperature:Ambient
- Immunogen:Human Serum Albumin
- Tested Applications:Fluoresence ELISA
- Purification:purified by immunoaffinity chromatography
- Cat. No.:RL600-402-033
- Supplier no.:600-402-033
Specifications
About this item
Recommended Dilutions:
Restore with deionized water (or equivalent); 1.0 ml
Type: Primary
Antigen: HSA
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human