Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
HistoZyme

HistoZyme

HistoZyme
Certificates
HistoZyme

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

HistoZyme is a proteolytic enzyme solution that results in superior staining over traditionally used enzyme pretreatments, such as pepsin and trypsin.

HistoZyme requires only a brief incubation at room temperature. Performing antigen recovery with HistoZyme, will allow you to dilute the primary antibody 5 to 10 fold further and achieve optimal staining.

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 95041-474
  • 95041-476
  • Enzyme Name:
  • HistoZyme
  • HistoZyme

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 15 ml
    Log in for availability
    Log in for availability
    Each (15ml)
    Each (15ml) - $235.36
    $235.36
    Each (15ml)
       $235.36

    Supplier Number: K046
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    95041-474
    Enzyme Name
    HistoZyme
  • 50 ml
    Log in for availability
    Log in for availability
    Each (50ml)
    Each (50ml) - $668.89
    $668.89
    Each (50ml)
       $668.89

    Supplier Number: K046-50
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    95041-476
    Enzyme Name
    HistoZyme

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.