Anti-IgG Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # RL713-402-002
Supplier: Rockland Immunochemical
Specifications
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Sheep
- Size:500 µL
- Epitope:Heavy and Light chain
- Cross Adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Antigen Synonyms:F(ab')2 Anti-Sheep IgG (H&L)
- Antibody Fragment:F(ab')2
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
- Storage Temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping Temperature:Ambient
- Immunogen:Sheep IgG whole molecule
- Tested Applications:Fluoresence ELISA, Fluoresence Western Blot
- Purification:purified by immunoaffinity chromatography
- Cat. No.:RL713-402-002
- Supplier no.:713-402-002
Specifications
About this item
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 500 µl
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Sheep