Order Entry
United States
ContactUsLinkComponent
96-Place Microcentrifuge Tube Rack
96-Place Microcentrifuge Tube Rack
Catalog # 470149-570
Supplier:  Ward's Science
undefined
96-Place Microcentrifuge Tube Rack
Catalog # 470149-570
Supplier:  Ward's Science
Supplier Number:  241014

Specifications

  • Description:
    96-Place microcentrifuge tube rack
  • Environmentally Preferable:
  • Cat. No.:
    470149-570
  • Supplier no.:
    241014

Specifications

About this item

High-volume rack

This rack is designed with wide spacing to allow storage of ninety six 1.5 or 2 ml tubes.The bright racks are designed for easy storage of microcentrifuge tubes. There are 11 mm diameter holes on one side and 9 mm on the other.

Ordering information: Color may vary from image.