Specifications
- Description:Water Condition Aqua Clear 100 mL
- Cat. No.:13197-144
- Supplier no.:170930000
Specifications
About this item
Protects water baths and other water systems against the growth of algae, bacteria, molds and biofilms.
- Permits longer use of water
- Does not harm film
- Helps prevent tubing from clogging
Protects pumps and thermostats and helps prevent tubing from clogging
Just 2 mL per liter of water adds a blue tint which will fade after about 3 months reminding you to change the water and add new conditioner.
Also permits longer use of water used in developer for color or X-ray films and will not harm film.