Hand Sink, Pedestal Base, Pedal Valve, Eagle MHC
Catalog # 71001-946
Supplier: Eagle Group
Specifications
- Bowl dimensions:9¾×13½×6¾"
- Cat. No.:71001-946
- Supplier no.:HSA-10-FA-1P
Specifications
About this item
Hand Sink is made up of Type 304 stainless steel.
- Heavy gauge
It features pedestal base with splash mounted faucet, single foot pedal valve, P-trap, tail piece and basket drain.