Hand Sink, Touch-Free System, Eagle MHC
Catalog # 89207-508
Supplier: Eagle Group
Specifications
- Description:Hand sink with waste receptacle
- Cat. No.:89207-508
- Supplier no.:HFL-5000
Specifications
About this item
It features deep-drawn positive drain sink bowl, basket drain, inverted 'V' edge to prevent spillage, removable stainless-steel trash container, and MICROGARD® antimicrobial protection.
- Other systems available features hand sink with skirt only (no waste receptacle) or hand sink with end splashes
- Hand sink is made up of heavy gauge type 304 stainless steel
Electronic-eye faucet with hydro-generator power source, hard roll towel dispenser with SaniTouch® mechanical touchless technology for one-at-a-time dispensing, Purell/LTX hands-free hand sanitizer dispenser, Gojo/TFX hands-free soap dispenser, FoodHandler glove rack and white bristle nail brush and Tucel soft bristle nail brush with holder.
Other systems available features hand sink with skirt only (no waste receptacle) or hand sink with end splashes.