Anti-IgG3 Mouse Monoclonal Antibody [clone: HP6050]
89366-042
: Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Immunoglobulin G3
- Antigen Symbol:IgG3
- Clonality:Monoclonal
- Clone:HP6050
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 μg
- Epitope:Hinge Region
- Gene ID:3502
- Antigen Synonyms:IgG3
- Storage Buffer:TRIS buffered saline containing 0.09% Sodium Azide (NaN3)
- Storage Temperature:Store at +4 °C or at –20 °C if preferred. This product should be stored undiluted. Storage in frost free freezers is not recommended. Should this product contain a precipitate we recommend microcentrifugation before use.
- Immunogen:Pool of 5-6 individually purified Human IgG3 myeloma proteins
- Tested Applications:Fluorescence-Activated Cell Sorting
- Purification:Purified IgG prepared by affinity chromatography on Protein A
- Cat. No.:89366-042
- Supplier no.:GTX76255
Mouse Monoclonal antibody [HP6050] to Human IgG3 (hinge region)
Type: Primary
Antigen: IgG3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope: Hinge Region
Host: Mouse
Isotype: IgG1
Reactivity: Human