Anti-CMA Mouse Monoclonal Antibody [clone: CC1]
89353-018
: Genetex
- Antibody Type:Primary
- Antigen Name:Chymase 1
- Antigen Symbol:CMA
- Clonality:Monoclonal
- Clone:CC1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:500 μl
- Gene ID:1215
- Antigen Synonyms:heart|mast cell|mast cell protease I|chymase|chymase 1 preproprotein transcript E|chymase 1 preproprotein transcript I|alpha-chymase
- Storage Buffer:10mM Phosphate-buffered salinie, pH 7.4, containing 0.2% BSA and 0.1% sodium azide
- Storage Temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Concentration:0.2 mg/ml
- Immunogen:Purified Human Skin Chymase
- Tested Applications:IHC-P
- Purification:Protein A affinity purified
- Cat. No.:89353-018
- Supplier no.:GTX72388
Mouse monoclonal [CC1] to Mast Cell Chymase
For IHC-P: Use at an assay dependent dilution. For WB: Use at a dilution of 1:200. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: CMA
Clonality: Monoclonal
Clone: CC1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: