To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Vimentin
- Antigen Symbol:VIM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:7 ml
- Gene ID:7431
- Antigen Synonyms:CTRCT30|HEL113
- Storage Buffer:10mM PO4, 200mM NaCl, pH 7.4, containing 1% BSA and 0.05% sodium azide
- Storage Temperature:Keep as concentrated solution, aliquot and store at 2-8°C. Do not freeze.
- Concentration:10µg/ml
- Immunogen:Purified human Vimentin
- Tested Applications:ICC/IF, IHC-P, IHC-Fr, WB
- Purification:Protein A purified
- Cat. No.:89352-490
- Supplier no.:GTX28545
Rabbit polyclonal antibody to Vimentin
Immunohistochemistry (IHC) 1:50-1:100 using polymer system or streptavidin-biotin system. Formalin-fixed paraffin-embedded (FFPE) tissue sections and cell smears do not require antigen retrieval.
Type: Primary
Antigen: VIM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human