Anti-VTN Mouse Monoclonal Antibody [clone: BV1]
Catalog # 89352-248
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Vitronectin
- Antigen Symbol:VTN
- Clonality:Monoclonal
- Clone:BV1
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:50 μg
- Gene ID:7448
- Antigen Synonyms:S Protein|Epibolin|Somatomedin B|VTN|Complement S Protein|Serum spreading factor|V75
- Storage Buffer:Phosphate buffered saline containing 0.2% BSA and 0.1% sodium azide
- Storage Temperature:Keep as concentrated solution, aliquot and store at 4°C
- Concentration:0.1 mg/ml
- Tested Applications:Fluorescence-Activated Cell Sorting, Immuno Assay, Purification
- Purification:IgG fraction
- Cat. No.:89352-248
- Supplier no.:GTX27165
Specifications
About this item
Mouse monoclonal [BV1] to Vitronectin
For FACS, IA, IP, P and WB: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: VTN
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human