Specifications
- Antibody Type:Primary
- Antigen Name:Pan Cytokeratin
- Antigen Symbol:p-CK
- Clonality:Monoclonal
- Clone:PCK-26
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Bovine
- Western Blot:Yes
- Size:100 µL
- Format:Ascites
- Antigen Synonyms:19|wide spectrum Cytokeratin|[cytokeratins 13|P-CK|24].|Cytokeratins|17|pan-cytokeratin|14|pan CK|16|pan-CK
- Storage Buffer:Ascitic fluid with 0.1% sodium azide
- Storage Temperature:–20 °C
- Immunogen:Full length native protein (purified) (Human epidermis).
- Tested Applications:Dot Blot
- Purification:Ascites
- Cat. No.:89361-912
- Supplier no.:GTX26401
Specifications
About this item
Cytokeratins, a group comprising at least 29 different proteins, are characteristic of epithelial and trichocytic cells. Cytokeratin 5, 6 and 8 are members of the type II neutral-to-basic subfamily. Monoclonal antibodies to cytokeratins are specific markers of epithelial cell differentiation and have been widely used as tools in tumor identification and classification. PCK-26 facilitates the typing of normal, metaplastic and neoplastic cells and it may aid in the discrimination of carcinomas and non-epithelial tumors such as sarcomas, lymphomas and neural tumors. It is also useful in detecting micro-metastases in lymph nodes and other tissues and for determining the origin of poorly differentiated tumors (1,2).
Type: Primary
Antigen: p-CK
Clonality: Monoclonal
Clone: PCK-26
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine