Anti-ALYREF Mouse Monoclonal Antibody [clone: 11G5]
Catalog # 89361-840
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Aly/REF export factor
- Antigen Symbol:ALYREF
- Clonality:Monoclonal
- Clone:11G5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:50 μl
- Gene ID:10189
- Antigen Synonyms:ALY|REF|ALY/REF|Aly/REF export factor|THOC4|BEF
- Storage Buffer:Preservative: 0.1% Sodium Azide; Constituents: PBS
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:2 mg/ml
- Immunogen:Recombinant human ALY / REF fusion protein.
- Tested Applications:ELISA, ICC/IF, IP, WB
- Purification:Protein A purified
- Cat. No.:89361-840
- Supplier no.:GTX26141
Specifications
About this item
Mouse monoclonal [11G5] to Aly
ELISA: Use at an assay dependent dilution. IF: Use at an assay dependent dilution. IP: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Detects a band of approximately 32 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: ALYREF
Clonality: Monoclonal
Clone: 11G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human