Anti-Nfatc2 Rabbit Polyclonal Antibody
Catalog # 89361-600
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:NFAT1
- Antigen Symbol:Nfatc2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:50 μl
- Gene ID:18019
- Modification:phospho-Ser54
- Storage Buffer:Phosphate-buffered saline, pH 7.3 containing 50% glycerol, 1.0 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptide - derived from a region of mouse NFAT 1 that contains serine 54
- Tested Applications:WB
- Purification:Immunogen affinity purified
- Cat. No.:89361-600
- Supplier no.:GTX25246
Specifications
About this item
Rabbit polyclonal to NFAT1 ( phospho S54 )
For WB: Use at a dilution of 1:1000. Predicted molecular weight: 154 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Nfatc2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse