Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-PSP Mouse Monoclonal Antibody [clone: YPSP-1]
  89360-550
 :  Genetex
Anti-PSP Mouse Monoclonal Antibody [clone: YPSP-1]
  89360-550
 :  Genetex
 :  GTX19070

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Prostate Secretory Protein
  • Antigen Symbol:
    PSP
  • Clonality:
    Monoclonal
  • Clone:
    YPSP-1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    250μg
  • Gene ID:
    4477
  • Antigen Synonyms:
    Microseminoprotein|beta-
  • Storage Buffer:
    Preservative: None. Constituents: 0.01M PBS. pH 7.2
  • Storage Temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Full length protein (purified) (Human).
  • Tested Applications:
    ELISA, WB
  • Purification:
    Protein G purified
  • Cat. No.:
    89360-550
  • Supplier no.:
    GTX19070

 

 

An abundant constituent of the human seminal plasma is a 14-kD protein of prostate origin known as beta-microseminoprotein, prostatic secretory protein (PRPS), Prostate secretory protein (PSP), or PSP94. It is a product of a single gene transcribed in the prostate but not in the testis. Specific receptors for this protein are found on spermatozoa and in the prostate.

ELISA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 14 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PSP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human