Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-MCSF Mouse Monoclonal Antibody [clone: 21]
  89360-490
 :  Genetex
Anti-MCSF Mouse Monoclonal Antibody [clone: 21]
  89360-490
 :  Genetex
 :  GTX18645

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Macrophage colony-stimulating factor 1
  • Antigen Symbol:
    MCSF
  • Clonality:
    Monoclonal
  • Clone:
    21
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Size:
    250μg
  • Gene ID:
    1435
  • Antigen Synonyms:
    MCSF|CSF-1|M-CSF|Lanimostim|Macrophage colony-stimulating factor 1|Processed macrophage colony-stimulating factor 1
  • Storage Buffer:
    Preservative: None. Constituents: 0.01M PBS. pH 7.2
  • Storage Temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Purified recombinant MCSF (Human).
  • Tested Applications:
    ELISA
  • Purification:
    Protein G purified
  • Cat. No.:
    89360-490
  • Supplier no.:
    GTX18645

 

 

Four distinct colony-stimulating factors (CSFs) that promote survival, proliferation and differentiation of bone marrow precursor cells have been well characterized: granulocyte macrophage CSF (GMCSF), granulocyte CSF (GCSF), macrophage CSF (MCSF), and Interleukin-3 (IL-3, Multi CSF). Both GMCSF and IL-3 are multipotential growth factors, stimulating proliferation of progenitor cells from more than one hematopoietic lineage. In contrast, GCSF and MCSF are lineage restricted hematopoietic growth factors, stimulating final mitotic divisions and the terminal cellular maturation of the partially differentiated hematopoietic progenitors. Macrophage CSF, also known as CSF1, is produced by monocytes, fibroblasts and endothelial cells. It stimulates the formation of macrophage colonies, enhances antibody-dependent, cell-mediated cytotoxicity by monocytes and macrophages, and inhibits bone resorption by osetoclasts. Natural human MCSF is a dimeric glycoprotein of 70-90 kD molecular weight, existing in multiple glycosylation forms. It binds to a 165 kD glycoprotein of the receptor tyrosine kinase subclass III, a family that includes the receptors for platelet derived growth factor (PDGF) and stem cell factor (SCF).

ELISA: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MCSF
Clonality: Monoclonal
Clone: 21
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human